Principaux documents
2633625
Supelco
Insert d'injection droit, splitless (non rempli)
pkg of 25 ea
About This Item
Code UNSPSC :
41115719
Conditionnement
pkg of 25 ea
L × D.E. × D.I.
95 mm × 5.0 mm × 3.4 mm
Description générale
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Shimadzu (17A) [with SPL-17 Injector]