Principaux documents
Z710865
Silicone membrane liner for Tunair shake flask
For 2.5 L flask
Synonyme(s) :
Silicone liner
About This Item
Code UNSPSC :
41122104
Nomenclature NACRES :
NB.22
Matériaux
polypropylene
Stérilité
non-sterile
Conditionnement
case of 5
Fabricant/nom de marque
IBI SS-3017
Description générale
Silicone membrane liner for Tunair shake flask consists of five 0.22 m silicone membrane cap linings. The filter lining in the cap assembly is replaced by tweaking the flanges of the inner-closure shell until they separate loose. Then it is pulled apart to remove the used lining. The lining can then be replaced by sandwiching it between two parts of the cap and the cap is snapped back together. While reassembling, ensure the flanges from the inner piece fits into the mated grooves in the outer piece. It is required for the cap assembly to stay intact while using.
Caractéristiques et avantages
  • Long-lasting unless punctured
  • Rinse with water only if visibly soiled.
  • Recommends autoclaving cap along with the filter.
Informations légales
Tunair is a trademark of IBI Scientific