Principaux documents
Z600768
Hellma® fluorescence cuvettes, semi Micro
High Performance Quartz Glass, spectral range 200-2,500 nm, pathlength 10x4 mm, chamber volume 1400 μL
Synonyme(s) :
Semi-Micro Fluorescence Cuvettes
About This Item
Code UNSPSC :
41121813
Nomenclature NACRES :
SB.32
Matériaux
Suprasil® quartz
Description
4 polished windows
Fabricant/nom de marque
Hellma 114F-QS
Paramètres
200-2,500 nm spectral range
H × l × P
46 mm × 12.5 mm × 12.5 mm
Volume de la chambre
1,400 μL
1400 μL
Longueur du trajet optique
10x4 mm
Description générale
with PTFE stopper
Conditionnement
Sold as packages containing a single cell (1EA = 1 cell)
Informations légales
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.