Z337269
BRAND® conical joint stopper, PP
joint: ST/NS 14/23
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.35
Produits recommandés
Joint
joint: ST/NS14/23
Caractéristiques
autoclavable
Fabricant/nom de marque
BRAND
Description générale
Hollow, with sealing ribs and square grip.
Autres remarques
BR144420 (India)
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG