Principaux documents
EP5820716002
Lid for Eppendorf(R) FA-45-6-30 Rotor
aerosol tight, aerosol tight, 5804 / 5804R / 5810 / 5810R, aluminum
About This Item
Code UNSPSC :
41103913
Nomenclature NACRES :
NB.82
Matériaux
aluminum
Caractéristiques
aerosol tight
Fabricant/nom de marque
Eppendorf® 5820716002 (Global)
Technique(s)
centrifugation: suitable
Compatibilité
for use with 5804 R
for use with 5804
for use with 5810 R
for use with 5810
Informations légales
Eppendorf is a registered trademark of Eppendorf AG