Principaux documents
Z556017
Sigma-Aldrich
Aldrich® aspirator bottle with tubulation
capacity 2 L, hose outlet O.D. 10 mm
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Capacité
2 L
D.E. de la sortie de tuyau
10 mm
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC