Principaux documents
Z530573
Sigma-Aldrich
Aldrich® reagent bottle
capacity 125 mL, joint: ST/NS 19/22, amber bottle
Synonyme(s) :
bottle reagent, reagent bottles
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
amber bottle
glass bottle
glass cap
round bottle
Joint
joint: ST/NS19/22
Caractéristiques
with cap
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Sigma-Aldrich
Capacité
125 mL
Description générale
Reagent bottles come with a ground glass hex head stopper. For light-sensitive samples, use the amber coated reagent bottles.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC