Principaux documents
Z146277
Scienceware® plastic-tubing cutter
jaw closes automatically when not in use
About This Item
Code UNSPSC :
41122406
Nomenclature NACRES :
NB.35
Fabricant/nom de marque
Bel-Art
Description générale
Cuts tubing of 1/16 to 3/4 in. O.D.
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP