Key Documents
Z150770
Glass-tubing cutter
About This Item
Code UNSPSC :
41122406
Nomenclature NACRES :
NB.35
Catégories apparentées
Description générale
Spring-loaded to stay open until used. Cuts tubing up to 1-1/2 in. (38 mm) diameter. Made of nickel-plated zinc-alloy die-casting.