Principaux documents
Z123358
Sigma-Aldrich
Presslok demountable cell holder
for window diameter 25 mm
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.32
Technique(s)
IR spectroscopy: suitable
Description générale
Assembly is accomplished by compressing cell plates between barrel assembly and self-locking cover. Highly efficient friction lock makes cell leak-proof. For use with all IR spectrometers. Pathlengths are varied by combinations of different PTFE spacers listed. Requires 25 or 32 mm diam. undrilled crystal windows listed (not included).
À utiliser avec