Z100528
Straight tube
joint: ST/NS 14/20
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Produits recommandés
Joint
joint: ST/NS14/20
Description générale
Provides a ST/NS: joint connection to single bulb ball-tube flasks and a hose connector to motor drive.