Principaux documents
WHA2017006
Whatman® Antibiotic Assay Discs
diam. 6 mm
Synonyme(s) :
Whatman paper, Z741310, microbial assays
About This Item
Code UNSPSC :
41111700
Nomenclature NACRES :
NB.79
Conditionnement
pkg of 1000 ea
Fabricant/nom de marque
Whatman 2017-006
Whatman Article No. 28418660 (US reference)
Diamètre
6 mm
Description générale
For determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
Caractéristiques et avantages
  • The antibiogram allows rational and selective chemotherapy.
  • Test discs can be coated with chemotherapeutic agents, placed on the innoculated nutrient agar and incubated.
  • The size of the inhibition zone is a measure for the effectiveness of the substances.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva