SYNC1324
Synthware™ chromatography flow control adapter
joint: ST/NS 24/40, with PTFE stopcock
Synonyme(s) :
Synthware C1324, Glass chromatography adapter
About This Item
Produits recommandés
Joint
joint: ST/NS24/40
Qualité
for chromatography
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C1324