Principaux documents
SYNC182403
Synthware chromatography reservoir
capacity 500 mL, joint: ST/NS 24/40
Synonyme(s) :
Synthware C182403, Glass chromatography reservoir
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Matériaux
colorless
glass
Joint
joint: ST/NS24/40
Qualité
for chromatography
Stérilité
non-sterile
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C182403
Capacité
500 mL
Adéquation
suitable for chromatography
Description générale
For flash chromatography.
Informations légales
Synthware is a trademark of Kemtech America Inc.