Key Documents
SYNA372440
Synthware Claisen adapter
joint: ST/NS 24/40
Synonyme(s) :
Synthware A372440, distillation adapter, glass Claisen adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A372440
Description générale
Provides dual entry into a variety of flasks.
Informations légales
Synthware is a trademark of Kemtech America Inc.