Principaux documents
DWK07-1200
Wheaton® Microliter μlplate Shallow Polypropylene
About This Item
Code UNSPSC :
41106200
Nomenclature NACRES :
NB.15
Matériaux:
conical bottom
polypropylene plate
round wells
Capacité:
350 μL
Matériaux
conical bottom
polypropylene plate
round wells
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
WHEATON® 07-1200
Capacité
350 μL
Application
Ideal for sample collection, storage, combinatorial chemistry and library applications
Caractéristiques et avantages
  • Premium virgin polypropylene shallow & medium μLplates is your assurance you achieve accurate results, the first time.
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.