Key Documents
BR91236
BRAND® glass beaker with spout, low form
volume 250 mL, with graduation
Synonyme(s) :
Beakers, Glass
About This Item
Code UNSPSC :
41121803
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass 3.3
Caractéristiques
with graduation
Fabricant/nom de marque
BRAND 91236
Volume
250 mL
Description générale
The BRAND® Glass Beaker with Spout is expertly crafted from Boro 3.3 glass, making it a reliable choice for laboratory applications. Designed in accordance with DIN EN ISO 3819 standards, this low-form beaker features clear graduation markings for precise measurement of liquids. The integrated spout allows for easy pouring, minimizing spills and enhancing accuracy during liquid transfer.
Autres remarques
Browse all glass beakers
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG