Key Documents
BR475800
BRAND® slide box
25 slides, 76 x 26 mm
Synonyme(s) :
Microscope slides, Microscopy, Slide box, Staining, Staining Accessories
About This Item
Code UNSPSC :
41123302
Nomenclature NACRES :
MA.01
Matériaux
polystyrene
Caractéristiques
Holds 25 x slides 76x26 mm
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 475800
Description générale
Sturdy design. Screw lid with dealing wire lug. Ideal for storing or shipping 5 thick or 10 thin slides. Slides protrude 10 mm above the box, to facilitate removal.
For slides, size (in, mm) 76 x 26. Numbered slots in base. Lid with index card.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Les clients ont également consulté
Slide 1 of 2
1 of 2