Key Documents
BR130254
BRAND® amber narrow-mouth bottle, LDPE, with screw cap
capacity 500 mL
Synonyme(s) :
Bottles, Plastics
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
bottle
low-density polyethylene cap
Joint
Joints threaded neck (GL25)
Caractéristiques
with cap
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 130254
Capacité
500 mL
Diam. × H
75 mm × 180 mm
Description générale
For storage of light-sensitive substances. With screw cap
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG