BR112805
BRAND® forcep with pointed ends
PMP, L 115 mm
Synonyme(s) :
Forceps
About This Item
Code UNSPSC :
41122405
Nomenclature NACRES :
NB.35
Produits recommandés
Matériaux
PMP
Caractéristiques
autoclavable at 121 C
Fabricant/nom de marque
BRAND 112805
Longueur
115 mm
Catégories apparentées
Description générale
The BRAND® Forceps with Pointed Ends are precision tools designed for handling delicate materials in laboratory settings. Made from high-quality PMP (polymethylpentene), these forceps are lightweight yet durable, making them ideal for various applications. Designed to withstand autoclaving at 121 °C, ensuring effective sterilization for safe use in sterile environments.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG