Key Documents
BAH369461010
SP Bel-Art® Sterileware Long Handled Sampling Spoon
white polystyrene, nominal capacity 14.79 mL , 3tsp, L × W 17.8 cm (7 in.) × 1.5625 in. (40 mm), sterile, pkg of 10 ea (individually wrapped)
Synonyme(s) :
Lab Scoop, dispensing tool, spoon
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Matériaux
white polystyrene
Stérilité
sterile
Conditionnement
pkg of 10 ea (individually wrapped)
Fabricant/nom de marque
Bel-Art H36946-1010
L × l
17.8 cm (7 in.) × 1.5625 in. (40 mm)
Capacité nominale
14.79 mL , 3tsp
Description générale
The long handle on these spoons enables users to reach into jars, bottles and other confined areas to minimize the potential of personal contact with sampled material.
Caractéristiques et avantages
  • Ten individually packaged spoons are sealed in an outer bag to create multiples layers of protection against contaminants and debris
  • Ideal for use in multi-zone clean rooms
  • FDA grade polystyrene safe for food, drug and cosmetic use
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP