Principaux documents
BAF131641419
Scienceware® biohazard disposal bags
width × H × thickness 14 in. (360 mm) × 19 in. (480 mm) × 0.04 mm (1.5 mil), pack of 200 ea, translucent red polypropylene
Synonyme(s) :
Scienceware® biohazard disposal bags
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Matériaux
translucent red polypropylene
Caractéristiques
label (With Biohazard Warning Label and Sterilization Indicator Patch)
Fits waste can 2.0 gallon
Conditionnement
pack of 200 ea
Fabricant/nom de marque
Bel-Art F13164-1419
Capacité
2.0 gal
7.6-15.1 L (2-4 gal)
width × H × thickness
14 in. (360 mm) × 19 in. (480 mm) × 0.04 mm (1.5 mil)
Description générale
Bright red polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
Caractéristiques et avantages
  • Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
  • High temperature steam autoclavable to 135°C (275°F)
  • Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
  • Made in USA
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP