Principaux documents
BAF131643848
Scienceware® biohazard disposal bags
width × H × thickness 970 mm (38 in.) × 48 in. (1220 mm) × 0.04 mm (1.5 mil), pack of 100 ea, red
Synonyme(s) :
biohaz waste bag, biohazard disposal bags, waste bag
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Matériaux
red
Conditionnement
pack of 100 ea
Fabricant/nom de marque
Bel-Art F13164-3848
Capacité
151.4-189.3 L (40-50 gal)
width × H × thickness
970 mm (38 in.) × 48 in. (1220 mm) × 0.04 mm (1.5 mil)
Description générale
Bright red polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
Caractéristiques et avantages
Bright red polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
  • Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
  • High temperature steam autoclavable to 135°C (275°F)
  • Pass ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests
  • Made in USA
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP