Key Documents
Z742374
Sigma-Aldrich
CANgas pressure regulator
Synonyme(s) :
Pressure regulator for gas
About This Item
Code UNSPSC :
40142201
Nomenclature NACRES :
NA.22
Niveau de qualité
Description générale
Aluminum pressure regulator with downstream metering valve and inlet pressure gauge with factory-set outlet pressure. Applicable for Messer® CANgas
Autres remarques
Product is currently only available to our customers in Europe
Informations légales
Messer is a registered trademark of Messer Group GmbH
Régulateur