Principaux documents
Z530670
Sigma-Aldrich
Aldrich® vacuum manifold with joints
positions, 4, joint: ST/NS 14/20
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.79
Joint
joint: ST/NS14/20
Caractéristiques
positions 4
Longueur
400 mm
Description générale
This practical design provides clearance to accommodate 250mL flasks with a snap of a Keck® clip. Manifolds are available with either <ST/>14/20, <ST/>24/40, or <ST/>29/32 joints. Glass stopcocks have a 4mm bore.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Keck is a registered trademark of Adcuram Lab Technologies GmbH