Principaux documents
502367
Vanne micrométrique (faible pression)
1/4-28 flangeless fittings
About This Item
Code UNSPSC :
41115854
eCl@ss :
32060101
Matériaux
PEEK
Description
micro-metering low pressure
Paramètres
800 psi max. pressure
Raccords
1/4-28 flangeless fittings
Description générale
Micro-Metering Valve is designed for the control of fluid flow rates. These needle valves are ideally used with peristaltic pumps, pump fluid-transfer, mass spectrometry and fraction collection.
Application
Micro-Metering valve (low pressure) can be used as a component of supercritical fluid extraction apparatus employing supercritical carbon dioxide, which may be used in the extraction and refining of soya-bean oil from full fat soya bean flakes and essential oil from the roots of Vetiveria zizanioides.
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Response surface method applied to supercritical carbon dioxide extraction of Vetiveria zizanioides essential oil.
Danh T L, et al.
Chemical Engineering Journal, 155(3), 617-626 (2009)
Characterization of soybean oil extracted by supercritical carbon dioxide and hexane.
Friedrich P J and List R G.
Journal of Agricultural and Food Chemistry, 30(1), 192-193 (1982)