Principaux documents
GN381080
Greiner CapMat for sealing multiwell plates
96 well, square nap, ethylene vinyl acetate, translucent
Synonyme(s) :
Greiner CapMats for sealing multiwell plates
About This Item
Code UNSPSC :
41122410
Conditionnement
pack of 50 ea
Fabricant/nom de marque
Greiner 381080
Disponibilité
available only in China
Résistance chim./phys.
Greiner CapMats are DMSO-resistant and withstand temperatures from -20°C to +60°C