Documents
CLS430165
Corning® tissue-culture treated culture dishes
D × H 35 mm × 10 mm
Synonyme(s) :
petri dish
About This Item
Code UNSPSC :
41121812
Nomenclature NACRES :
NB.13
Matériaux
polystyrene
round
Stérilité
sterile
Conditionnement
sleeve of 20
Fabricant/nom de marque
Corning 430165
P × H
35 mm × 10 mm
Superficie
8 cm2
Caractéristiques et avantages
  • Manufactured from optically clear virgin polystyrene
  • Treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Lid is supplied with vents to provide consistent gas exchange
Informations légales
Corning is a registered trademark of Corning, Inc.