Documents
CLS430293
Corning® tissue-culture treated culture dishes
D × H 100 mm × 20 mm
Synonyme(s) :
petri dish
About This Item
Code UNSPSC :
41121812
Nomenclature NACRES :
NB.13
Matériaux
polystyrene
round
Stérilité
sterile
Utilisation
11.0-16.5 mL Optimal culture medium volume range (Optimal culture medium volume)
Conditionnement
sleeve of 10
Fabricant/nom de marque
Corning 430293
P × H
100 mm × 20 mm
Superficie
55 cm2
Caractéristiques et avantages
  • Manufactured from optically clear virgin polystyrene
  • Treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Lid is supplied with vents to provide consistent gas exchange
Informations légales
Corning is a registered trademark of Corning, Inc.