CLS3368
Microplaque Corning® 96 puits pour tests EIA/RIA
flat bottom clear, polystyrene, bag of 25, sterile, lid: no
Synonyme(s) :
96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 welll plate, EIA/RIA plate, immunoassay plate
475,00 $
About This Item
475,00 $
Produits recommandés
Matériaux
flat bottom clear
polystyrene
polystyrene
round wells
Stérilité
sterile
Caractéristiques
lid: no
skirt
plate format: 96 well Easy-Wash™
well: flat bottom, clear
Conditionnement
case of 100
bag of 25
Fabricant/nom de marque
Corning 3368
Taille
96 wells , 96 well plate
Volume des puits
330-360 μL , Note: CLS3366 and CLS3797 have well volume capacities of 330 uL
Volume de travail par puits
75-200 μL
Adéquation
suitable for (EIA/RIA; immunoassays)
Type de liaison
non-treated surface
Catégories apparentées
Description générale
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
- Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
- Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm2
- Easy Wash design (round to narrowed flat well bottom) for improved washing in immunoassays
- EIA/RIA plates are tested on lot-to-lot basis and the certification results for each lot are available upon request
- Recommended working volume of up to 75 to 200 μL
- Conforms to standard microplate footprint and dimensions