Z615617
Julabo® bath carrier trays for SW22/SW23
to fit 15 Erlenmeyer flasks 200 cc
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.77
Produits recommandés
Fabricant/nom de marque
Julabo 8970363
Julabo
Catégories apparentées
Description générale
Erlenmeyer flasks can be positioned in the shaking water bath using the stainless steel tray. A maximum of 15 Erlenmeyer flasks with 200 ml capacity fit on a standard tray.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.