Principaux documents
EPM1289-0600
Test Tube Rack, for Eppendorf® New Brunswick Innova® and Eppendorf® Excella® Shakers
holds 20 x 50 mL conical tubes (26-30 mm diam.)
1 EA
587,00 $
587,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
587,00 $
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.73
587,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
holds 20 x 50 mL conical tubes (26-30 mm diam.)
Fabricant/nom de marque
Eppendorf® M1289-0600 (Global)
Taille
450x80x129 mm
Description générale
Test Tube Racks have an adjustable angle and tube height ­support. Made from stainless-steel with inserts to help retain tubes and ­prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc. 
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG