Z509337
Weighing funnel
size S, anti-static PP
Synonyme(s) :
Weighing Apparatus, Weighing Device
About This Item
Produits recommandés
Matériaux
anti-static PP
Taille
S
Conditionnement
pkg of 50 ea
Catégories apparentées
Description générale
- Sit flat on balance - No rolling, no spilling!
- Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer
- Residual sample can be rinsed to guarantee 100% transfer
- Suitable for safe handling of hazardous materials
- Available in five different sizes: extra small, small, medium, large, and extra large