The material can withstand a standard autoclave cycle (steam 30 minutes at 15 psi, 250° F (121° C)), but for repeated sterilization, this product would not be suitable.
Z280372
PVC laboratory tubing
I.D. × O.D. 1/4 in. × 3/8 in.
Synonyme(s) :
Saint Gobain Versilon ABH02017
62,20 $
About This Item
62,20 $
Produits recommandés
Conditionnement
pkg of 100 feet
D.I. × D.E.
1/4 in. × 3/8 in.
Taille de la paroi
1/16 in.
Les clients ont également consulté
-
Is this tubing autoclavable?
1 answer-
Helpful?
-
-
Could you provide information about the maximum psi of PVC laboratory tubing Z280372-1PAK?
1 answer-
The maximum operating pressure is 30 pounds per square inch at 73 degrees Fahrenheit.