Principaux documents
BR143265
BRAND® special laboratory tubing
I.D. × O.D. × W 5 mm × 8 mm × 1.5 mm, L 20 m
Synonyme(s) :
Special laboratory tubing, Tubing
About This Item
Code UNSPSC :
41105108
Nomenclature NACRES :
NB.35
Matériaux
PVC
Fabricant/nom de marque
BRAND 143265
D.I. × D.E. × l
5 mm × 8 mm × 1.5 mm
Longueur
20 m
Description générale
Particularly high-grade material, manufactured to a first-class standard. Good flexibility and chemical resistance make this tubing ideal for use with apparatus assemblies. Autoclavable (121 °C).
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG