Principaux documents
Z255815
Sigma-Aldrich
Latex rubber laboratory tubing
I.D. × O.D. 1/4 in. × 3/8 in.
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.35
Description
standard
Conditionnement
pkg of 100 feet
D.I. × D.E.
1/4 in. × 3/8 in.
Taille de la paroi
1/16 in.
Description générale
High-purity, seamless, amber tubing for general-purpose use with liquids and gases. Durometer: 35 ±5. Talc-free.