Principaux documents
Z111392
Sigma-Aldrich
Heavy-duty Wig-L-Bug® grinder/mixer analog
AC/DC input 115 V AC
About This Item
Code UNSPSC :
12000000
Le produit Z111392 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Disponibilité
not available in EU
Description générale
Useful for mixing and grinding that require constant and long periods of operation. Has a 30 minute timer and fan-cooled motor. Comes with vial adapter for 1/2 in. diam. × 1 in. long vials.
Informations légales
Wig-L-Bug is a registered trademark of Dentsply Rinn