Documents
Z111554
Sigma-Aldrich
Steel vial and ball pestle for analog Wig-L-Bug® grinder/mixer
stainless steel, diam. × L 1/2 in. × 1.0 in.
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.32
Matériaux
stainless steel
Disponibilité
not available in EU
Technique(s)
IR spectroscopy: suitable
Diam. × L
1/2 in. × 1.0 in.
Description générale
For grinding and mixing hard materials. Includes 1/4 in. diam. ball pestle.
Informations légales
Wig-L-Bug is a registered trademark of Dentsply Rinn