Principaux documents
Z529656
Sigma-Aldrich
Digital Wig-L-Bug® grinder/mixer
low profile
About This Item
Code UNSPSC :
41121813
Entrée CA/CC
110 V
Disponibilité
not available in EU
Larg. × P × H
11 1/2 in. × 8 in. × 4 1/2 in.
Description générale
  • Easy-to-use icon-driven control panel
  • Membrane keypad is easily cleaned and disinfected
  • Safety cover shields capsules, reduces noise
  • Speeds of 3,800, 4,200, 4,600, and 4,800 rpm
  • Accurate digital circuitry with LED readout of time and speed
  • Vials and pestles sold separately
Informations légales
Wig-L-Bug is a registered trademark of Dentsply Rinn