Principaux documents
TEM-G200CP
Sigma-Aldrich
Glider Grids Standard Copper/Palladium Square Mesh
size 200 mesh, vial of 100
Synonyme(s) :
SQUARE MESH CU/PD 100 VIAL
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
vial of 100
Taille
200 mesh
Description générale
Gilder Grids Standard Square Mesh: Copper/Palladium Grid, 200 mesh, 3.05mm diameter, 0.7 mil (18μm) thickness. Pitch 125μm, Hole 90μm, Bar 35μm