930318
TEM Grids
Formvar Supported Copper Square Mesh, grid size 200 mesh, box of 25
Synonyme(s) :
TEM grids
About This Item
Produits recommandés
product name
Formvar Supported Copper Square Mesh, grid size 200 mesh, box of 25
Matériaux
copper grid
formvar film
Description
film layer thickness 10 nm
Conditionnement
box of 25
Taille de la grille
200 mesh