Documents
SLW2115/04M
Pyrex® Weighing scoop
6 mL
Synonyme(s) :
Pyrex, SciLabware 2115/04M
About This Item
Code UNSPSC :
41121812
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
Conditionnement
pack of 10 ea
Fabricant/nom de marque
SciLabware 2115/04M
L totale
85 mm
Capacité
6 mL
Description générale
Glass scoop
  • With hollow stem to aid introduction of the powder into narrow neck vessels e.g. volumetric flasks
  • The flat base ensures stability on the balance pan
Informations légales
PYREX is a registered trademark of Corning, Inc.