Principaux documents
CLS998590
Pyrex® watch glasses
O.D. 90 mm
Synonyme(s) :
Corning® 9985
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Corning 9985-90
D.E.
90 mm
Catégories apparentées
Description générale
Heavy-wall, borosilicate glass with fire-polished edge provides durability. May be slightly out-of-round due to O.D. variability.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.