Documents
BR452215
BRAND® Inoculation loop with needle, disposable
volume 1+10 μL, yellow
Synonyme(s) :
Inoculation loops
About This Item
Code UNSPSC :
42142523
Nomenclature NACRES :
NB.22
Matériaux
polystyrene
yellow
Stérilité
sterile; γ-irradiated
Conditionnement
pack of 1000 ea
pkg of 8000 ea (400 bags of 20)
Fabricant/nom de marque
BRAND 452215
Volume
1+10 μL
Description générale
For inoculation of nutrient media. Available either with loop at one end and needle at the other or loops at both ends. High flexibility permits gentle streaking without damaging the nutrient surface. γ-ray sterilized.
Caractéristiques et avantages
• Sterile (SAL 10-6)• High flexibility for agar-saving smears• Usable at both ends
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Les clients ont également consulté
Slide 1 of 2
1 of 2