Skip to Content
MilliporeSigma
All Photos(1)

Documents

55058

Bottom-of-the-Bottle filter/sparger

pore size 2 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

packaging

pkg of 1 ea

manufacturer/tradename

Upchurch Scientific, Inc A-435

diam.

28 mm

pore size

2 μm

General description

Bottom-of-the-Bottle filter/sparger is an accessory for use with Mobile Phase Reservoir System.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Bottom-of-the-Bottle is a trademark of IDEX Health & Science, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service