Skip to Content
MilliporeSigma
All Photos(1)

Documents

56705

Inlet Filter Assembly

pore size 10 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

material

Tefzel® unit (fittings included)
polyethylene filter

description

UHMWPE Bottom-of-the-Bottle
solvent filter assembly

feature

replaceable frits

manufacturer/tradename

Upchurch Scientific, Inc A-426

pore size

10 μm

General description

10μm Inlet Filter - This metal-free Upchurch unit consists of a replaceable 10μm ultra-high molecular weight polyethylene filter that screws into a Tefzel holder. The unit connects to the inlet tubing through a flangeless Tefzel fitting (included).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Bottom-of-the-Bottle is a trademark of IDEX Health & Science, LLC
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company

replacement part


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service