Documents
55062
Cap for Mobile Phase Reservoir System
GL 45 bottle cap,
number of holes 3
About This Item
UNSPSC Code:
41115700
material
red polypropylene cap
white Tefzel® insert
packaging
pkg of 1 ea
manufacturer/tradename
Upchurch Scientific, Inc A-620
fitting
GL 45 bottle cap
Features and Benefits
Cap fits 1-liter or 2-liter reservoir system bottles. Cap insert has two holes for 1/8 in. tubing, one for luer taper
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)