Key Documents
22999
Supelco
Valco® Unions
internal reducing, bore 0.75 mm, for connecting 1/8 to 1/16 in.
Synonym(s):
Internal reducing union
Select a Size
1 EA
$62.10
$62.10
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$62.10
About This Item
UNSPSC Code:
41115700
$62.10
Please contact Customer Service for Availability
Request a Bulk Order
material
stainless steel (nuts and ferrules included)
packaging
pkg of 1 ea
bore
0.75 mm
fitting
1/16 in. Valco
1/8 in. Valco
compatibility
for connecting 1/8 to 1/16 in.
General description
Stainless steel, unless noted otherwise. Nuts and ferrules included.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)