Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK181

Sigma-Aldrich

High Sensitivity Glucose Assay Kit

Sufficient for 100 Fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12161503
NACRES:
NA.84

detection method

fluorometric

relevant disease(s)

endocrinological disorders, diabetes; cancer

storage temp.

−20°C

General description

Glucose is a primary energy source that naturally occurs in its free state in fruits and other plant parts. Abnormal glucose levels have been associated with several metabolic dysfunctions such as hypoglycemia, hyperglycemia, and diabetes mellitus. Measurements of glucose levels in tissues and body fluids (such as blood and urine) are often used for the diagnosis of glucose-related disorders. Glucose levels are also monitored to check the efficacy of therapeutics such as insulin and sulphonylureas in type 2 diabetics.

Suitability

High Sensitivity Glucose Assay Kit is a highly sensitive assay for determining glucose levels (ranging from 20-100pmole/well) in a variety of biological samples.

Principle

Glucose concentration is determined by a coupled enzyme assay, which results in a fluorometric (λex = 535/λem = 587 nm) product proportional to the glucose present.

Kit Components Only

Product No.
Description

  • Glucose Assay Buffer

  • Glucose Probe, in DMSO

  • Glucose Enzyme Mix

  • Glucose Substrate Mix

  • Glucose Standard, 100 mM

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diabetes mellitus and oxidative stress?a concise review.
Asmat U, et al.
Saudi Pharmaceutical Journal : SPJ : The Official Publication of the Saudi Pharmaceutical Society, 24(5), 547-553 (2016)
Berg J M, et al.
Biochemistry (2002)
Management of type-2 diabetes mellitus in adults: focus on individualizing non-insulin therapies.
Brunetti L and Julie K
P & T : A Peer-Reviewed Journal for Formulary Management, 97(12), 687-687 (2012)
Diagnosis and classification of diabetes mellitus.
Diabetes Care, 33(1), S62-S62 null
Li-Li Zhu et al.
International journal of biological sciences, 17(1), 107-118 (2021-01-05)
Aerobic glycolysis, also known as the Warburg effect, is emerged as a hallmark of most cancer cells. Increased aerobic glycolysis is closely associated with tumor aggressiveness and predicts a poor prognosis. Pancreatic ductal adenocarcinoma (PDAC) is characterized by prominent genomic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service