Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAK476

Sigma-Aldrich

Glucose Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Glucose Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.84

Quality Level

application(s)

cosmetics
food and beverages

detection method

colorimetric
fluorometric

relevant disease(s)

diabetes; metabolic disorders; endocrinological disorders; cancer

shipped in

wet ice

storage temp.

−20°C

General description

Glucose (C6H12O6) is a key diagnostic parameter for many metabolic disorders. Increased glucose levels have been associated with diabetes mellitus, hyperactivity of the thyroid, pituitary, and adrenal glands. Decreased levels are found in insulin secreting tumors, myxedema, hypopituitarism and hypoadrenalism.

Features and Benefits

  • The linear detection range of glucose for the colorimetric assay is 5 to 300 µM (90 µg/dL -5.4 mg/dL) and 1 to 30 µM for the fluorometric assay.
  • Compatible with high-throughput handling systems.
  • The procedure involves addition of a single working reagent and incubation for 30 minutes at room temperature.

Suitability

The kit is suitable for glucose determination in serum, plasma, milk, culture medium, food, beverages, and other biological preparations

Principle

The Glucose Assay Kit uses a single Working Reagent that combines the glucose oxidase reaction and color reaction in one step. The color intensity of the reaction product at 570 nm or fluorescence intensity at λEx = 530 nm/λEm = 585 is directly proportional to glucose concentration in the sample.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service